siRNA Details
Gene Symbol: LCE3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032563.1 | NP_115952.1 | 2 | 379 | ||
GenBank mRNA | AB048288.1 | - | 2 | 379 | ||
BC101784.1 | - | 2 | 362 | 8069290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
