siRNA Details
Gene Symbol: TXLNGY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045128.1 | 8, 9 | 1174 | |||
GenBank mRNA | AF332225.1 | - | 2, 3 | 450 | ||
AL832583.2 | - | 3, 4 | 4720 | |||
CD242693.1 | - | 3, 4 | 517 |
