siRNA Details
Gene Symbol: LYZL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032517.4 | NP_115906.3 | 2 | 194 | ||
XM_005252627.3 | XP_005252684.1 | 2 | 204 | |||
XM_017016791.1 | XP_016872280.1 | 2 | 188 | |||
GenBank mRNA | AY358694.1 | - | 2 | 139 | ||
BC021730.2 | - | 2 | 160 | 4825273 | ||
BP369317.1 | - | 2 | 205 |
Gene Symbol: LYZL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183058.2 | NP_898881.2 | 2 | 150 | ||
XM_011519306.2 | XP_011517608.1 | 2 | 193 | |||
XM_011519307.2 | XP_011517609.1 | 2 | 193 | |||
GenBank mRNA | AF139543.1 | - | 2 | 129 | ||
BC066294.1 | - | 2 | 150 | 4374284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
