siRNA Details
Gene Symbol: EBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278636.1 | NP_001265565.1 | 5 | 900 | ||
NM_032565.4 | NP_115954.1 | 4 | 838 | |||
NR_103802.1 | 5 | 959 | ||||
NR_103803.1 | 3 | 768 | ||||
GenBank mRNA | AF243433.1 | - | 4 | 827 | ||
BC021021.1 | - | 5 | 877 | |||
BC073152.1 | - | 5 | 959 | |||
BC092471.1 | - | 4 | 833 | 4796715 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
