siRNA Details
Gene Symbol: LINC00852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026829.1 | 1 | 701 | |||
GenBank mRNA | AF280797.2 | - | 1 | 701 | ||
BC015602.2 | - | 1 | 653 | |||
BC132802.1 | - | 1 | 330 | |||
BC132804.1 | - | 1 | 330 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
