siRNA Details
Gene Symbol: MYPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256267.1 | NP_001243196.1 | 14, 15 | 3120 | ||
NM_001256268.1 | NP_001243197.1 | 17, 18 | 3529 | |||
NM_032578.3 | NP_115967.2 | 13, 14 | 3225 | |||
NR_045662.3 | 13, 14 | 2360 | ||||
NR_045663.3 | 11, 12 | 3062 | ||||
XM_017016833.1 | XP_016872322.1 | 14, 15 | 3561 | |||
XM_017016834.1 | XP_016872323.1 | 13, 14 | 3019 | |||
GenBank mRNA | AF328296.1 | - | 13, 14 | 3420 | ||
AL832002.2 | - | 13, 14 | 3148 | |||
AL832379.1 | - | 13, 14 | 2322 | |||
AL834247.2 | - | 13, 14 | 3147 | |||
BC144334.1 | - | 11, 12 | 2824 | |||
BX647637.1 | - | 13, 14 | 3156 | |||
BX647640.1 | - | 14, 15 | 3113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
