siRNA Details
Gene Symbol: DLGAP1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_119377.1 | 1 | 224 | |||
GenBank mRNA | AK312196.1 | - | 1 | 317 | ||
BC094703.1 | - | 1 | 223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
