siRNA Details
Gene Symbol: BTRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256856.1 | NP_001243785.1 | 14 | 3145 | ||
NM_003939.4 | NP_003930.1 | 14 | 3115 | |||
NM_033637.3 | NP_378663.1 | 15 | 3223 | |||
XM_006718054.2 | XP_006718117.1 | 15 | 3523 | |||
XM_011540320.2 | XP_011538622.1 | 12 | 2959 | |||
XM_017016870.1 | XP_016872359.1 | 14 | 3445 | |||
XM_017016871.1 | XP_016872360.1 | 15 | 3114 | |||
XM_017016872.1 | XP_016872361.1 | 15 | 3286 | |||
XM_017016873.1 | XP_016872362.1 | 15 | 3142 | |||
XM_017016874.1 | XP_016872363.1 | 13 | 3037 | |||
GenBank mRNA | BC006204.2 | - | 1 | 185 | ||
CR749295.1 | - | 14 | 3076 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
