siRNA Details
Gene Symbol: MDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145337.2 | NP_001138809.1 | 11 | 3626 | ||
NM_001145339.2 | NP_001138811.1 | 9 | 3852 | |||
NM_001145340.2 | NP_001138812.1 | 5 | 3179 | |||
NM_001278462.1 | NP_001265391.1 | 6 | 3257 | |||
NM_002392.5 | NP_002383.2 | 11 | 4017 | |||
XM_005268872.4 | XP_005268929.1 | 11 | 3945 | |||
XM_006719399.3 | XP_006719462.1 | 9 | 3600 | |||
XM_006719400.3 | XP_006719463.1 | 10 | 3906 | |||
GenBank mRNA | BC065505.1 | - | 1 | 312 | ||
BC107414.1 | - | 1 | 561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
