siRNA Details
Gene Symbol: ZNF678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178549.3 | NP_848644.2 | 4 | 7825 | ||
NR_033184.1 | 4 | 7853 | ||||
GenBank mRNA | BC007286.1 | - | 1 | 497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
