siRNA Details
Gene Symbol: LINC00839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026827.1 | 3 | 698 | |||
GenBank mRNA | AK090789.1 | - | 3 | 645 | ||
AK092104.1 | - | 3 | 648 | |||
BC007394.2 | - | 3 | 651 | |||
BI517331.1 | - | 3 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
