siRNA Details
Gene Symbol: TBRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032811.2 | NP_116200.2 | 3 | 660 | ||
NR_016021.1 | 3 | 660 | ||||
XM_017018442.1 | XP_016873931.1 | 1 | 1618 | |||
GenBank mRNA | AK027527.1 | - | 3 | 631 | ||
AK074140.1 | - | 3 | 383 | |||
AK222936.1 | - | 3 | 636 | |||
AY696294.1 | - | 1 | 106 | |||
BC041043.1 | - | 3 | 645 | |||
BC064392.1 | - | 3 | 609 | |||
BC107428.1 | - | 3 | 650 | |||
DQ144542.1 | - | 3 | 422 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
