siRNA Details
Gene Symbol: ITIH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030569.6 | NP_085046.5 | 14 | 5263 | ||
NM_032817.5 | NP_116206.4 | 10 | 4598 | |||
XM_011519713.2 | XP_011518015.1 | 15 | 5338 | |||
GenBank mRNA | AK027547.1 | - | 1 | 1563 | ||
BC050649.1 | - | 1 | 924 | |||
BC069714.1 | - | 1 | 1474 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
