siRNA Details
Gene Symbol: BTRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256856.1 | NP_001243785.1 | 14 | 4607 | ||
NM_003939.4 | NP_003930.1 | 14 | 4577 | |||
NM_033637.3 | NP_378663.1 | 15 | 4685 | |||
XM_006718054.2 | XP_006718117.1 | 15 | 4985 | |||
XM_011540320.2 | XP_011538622.1 | 12 | 4421 | |||
XM_017016870.1 | XP_016872359.1 | 14 | 4907 | |||
XM_017016871.1 | XP_016872360.1 | 15 | 4576 | |||
XM_017016872.1 | XP_016872361.1 | 15 | 4748 | |||
XM_017016873.1 | XP_016872362.1 | 15 | 4604 | |||
XM_017016874.1 | XP_016872363.1 | 13 | 4499 | |||
GenBank mRNA | BC006204.2 | - | 1 | 1647 | ||
BQ188694.1 | - | 1 | 65 | |||
CR749295.1 | - | 14 | 4539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
