siRNA Details
Gene Symbol: TRIM52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032765.2 | NP_116154.1 | 1, 2 | 1126 | ||
GenBank mRNA | AK054802.1 | - | 1, 2 | 1103 | ||
BC007372.2 | - | 1, 2 | 1126 | 3636274 | ||
JX896146.1 | - | 1, 2 | 821 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
