siRNA Details
Gene Symbol: ATP1A1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024124.2 | 2 | 672 | |||
NR_024125.2 | 3 | 948 | ||||
NR_024126.1 | 3 | 711 | ||||
GenBank mRNA | BC007373.2 | - | 3 | 711 | ||
BC018892.2 | - | 3 | 711 | |||
BC098428.1 | - | 3 | 948 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
