siRNA Details
Gene Symbol: MINA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042533.2 | NP_001035998.1 | 10 | 1902 | ||
NM_001261829.1 | NP_001248758.1 | 10 | 1432 | |||
NM_032778.5 | NP_116167.3 | 10 | 1899 | |||
NM_153182.3 | NP_694822.2 | 10 | 1435 | |||
XM_005247838.4 | XP_005247895.1 | 10 | 1652 | |||
XM_011513244.2 | XP_011511546.1 | 6 | 1164 | |||
GenBank mRNA | AB083189.1 | - | 10 | 1538 | ||
AB083190.1 | - | 10 | 1535 | |||
AK027299.1 | - | 10 | 1382 | |||
AY302110.3 | - | 10 | 1605 | |||
AY390536.1 | - | 10 | 1587 | |||
AY456380.1 | - | 10 | 1169 | |||
BC014928.1 | - | 10 | 1589 | 3916148 | ||
CR627479.1 | - | 9 | 1188 | |||
DQ453796.1 | - | 10 | 1399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
