siRNA Details
Gene Symbol: ALG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032834.3 | NP_116223.3 | 3 | 919 | ||
GenBank mRNA | AJ312278.1 | - | 3 | 919 | ||
AK027657.1 | - | 1 | 157 | |||
AK292488.1 | - | 3 | 685 | |||
BC033730.1 | - | 3 | 700 | |||
BC070347.1 | - | 3 | 704 | 3839674 | ||
BC090948.1 | - | 3 | 686 | 5205402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
