siRNA Details
Gene Symbol: FAM104A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098832.1 | NP_001092302.1 | 4 | 640 | ||
NM_001289410.1 | NP_001276339.1 | 2 | 477 | |||
NM_001289411.1 | NP_001276340.1 | 3 | 495 | |||
NM_001289412.1 | NP_001276341.1 | 4 | 413 | |||
NM_032837.2 | NP_116226.2 | 3 | 577 | |||
GenBank mRNA | AK027681.1 | - | 3 | 462 | ||
AK074755.1 | - | 3 | 463 | |||
AK304556.1 | - | 3 | 550 | |||
BC011054.1 | - | 3 | 302 | 4177665 | ||
BC025238.1 | - | 3 | 340 | 4905931 | ||
BG773537.1 | - | 3 | 432 | |||
BX537858.1 | - | 4 | 544 | |||
BX648968.1 | - | 5 | 556 | |||
DB265439.1 | - | 2 | 477 | |||
HY026689.1 | - | 4 | 413 | |||
HY037907.1 | - | 4 | 398 | |||
U92989.1 | - | 4 | 384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
