siRNA Details
Gene Symbol: PTPN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002834.3 | NP_002825.3 | 16 | 4756 | ||
XM_006719526.2 | XP_006719589.1 | 16 | 4546 | |||
XM_011538613.2 | XP_011536915.1 | 16 | 4582 | |||
XM_017019722.1 | XP_016875211.1 | 16 | 4573 | |||
GenBank mRNA | AK312147.1 | - | 1 | 192 | ||
BC007869.2 | - | 1 | 245 | |||
BC025181.1 | - | 1 | 321 | |||
BC030949.1 | - | 1 | 265 | |||
BX537632.1 | - | 1 | 1845 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
