siRNA Details
Gene Symbol: BFSP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135276.1 | 4 | 542 | |||
NR_135277.1 | 3 | 427 | ||||
NR_135278.1 | 4 | 329 | ||||
GenBank mRNA | AA746278.1 | - | 2 | 138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
