siRNA Details
Gene Symbol: TCEAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006933.1 | NP_001006934.1 | 3 | 352 | ||
NM_032926.2 | NP_116315.1 | 3 | 340 | |||
GenBank mRNA | AF163263.1 | - | 3 | 289 | ||
BC007622.1 | - | 3 | 331 | 3355622 | ||
BC008703.1 | - | 3 | 340 | 2966921 |
Gene Symbol: TCEAL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012979.2 | NP_001012997.1 | 3 | 400 | ||
GenBank mRNA | BC132922.1 | - | 3 | 277 | 40146944 | |
BC132924.1 | - | 3 | 277 | 40146946 |
Gene Symbol: TCEAL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006938.2 | NP_001006939.2 | 3 | 251 | ||
GenBank mRNA | BC071675.1 | - | 3 | 234 | 5197831 | |
DN993514.1 | - | 3 | 363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
