siRNA Details
Gene Symbol: ZNF587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204817.1 | NP_001191746.1 | 3 | 3075 | ||
NM_032828.3 | NP_116217.1 | 3 | 3078 | |||
GenBank mRNA | AK027616.1 | - | 4 | 2719 | ||
AK160374.1 | - | 2 | 2954 |
