siRNA Details
Gene Symbol: ALG10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013620.3 | NP_001013642.1 | 3 | 795 | ||
XM_005268665.4 | XP_005268722.1 | 3 | 576 | |||
GenBank mRNA | AK307955.1 | - | 3 | 750 | ||
AY845858.1 | - | 3 | 656 | |||
BC137413.1 | - | 3 | 711 | 9021411 | ||
BC137414.1 | - | 3 | 711 | 9021412 | ||
BC144617.1 | - | 3 | 685 |
Gene Symbol: ALG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032834.3 | NP_116223.3 | 3 | 975 | ||
GenBank mRNA | AJ312278.1 | - | 3 | 975 | ||
AK027657.1 | - | 1 | 213 | |||
AK292488.1 | - | 3 | 741 | |||
BC033730.1 | - | 3 | 756 | |||
BC070347.1 | - | 3 | 760 | 3839674 | ||
BC090948.1 | - | 3 | 742 | 5205402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
