siRNA Details
Gene Symbol: AJUBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032876.5 | NP_116265.1 | 2 | 1414 | ||
GenBank mRNA | AY169959.1 | - | 2 | 1110 | ||
BC007580.1 | - | 2 | 1110 | 3139903 | ||
BC034968.2 | - | 2 | 1414 | 4837383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
