siRNA Details
Gene Symbol: WDR53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182627.1 | NP_872433.1 | 3, 4 | 959 | ||
GenBank mRNA | AK314105.1 | - | 3, 4 | 968 | ||
BC054030.1 | - | 3, 4 | 959 | 6145598 | ||
EF011620.1 | - | 3, 4 | 959 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
