siRNA Details
Gene Symbol: FAM83A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288587.2 | NP_001275516.1 | 4 | 1474 | ||
NM_032899.6 | NP_116288.2 | 5 | 4165 | |||
NM_207006.2 | NP_996889.1 | 6 | 3241 | |||
XM_005251087.2 | XP_005251144.1 | 3 | 3370 | |||
GenBank mRNA | AF497803.1 | - | 5 | 1481 | ||
BC007828.2 | - | 4 | 1145 | 3610547 | ||
BQ927762.1 | - | 1 | 235 | |||
DQ280322.1 | - | 4 | 1184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
