siRNA Details
Gene Symbol: TCEAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006933.1 | NP_001006934.1 | 3 | 366 | ||
NM_032926.2 | NP_116315.1 | 3 | 354 | |||
GenBank mRNA | AF163263.1 | - | 3 | 303 | ||
AK098494.1 | - | 3 | 297 | |||
BC007622.1 | - | 3 | 345 | 3355622 | ||
BC008703.1 | - | 3 | 354 | 2966921 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
