siRNA Details
Gene Symbol: ATG4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032852.3 | NP_116241.2 | 11 | 2029 | ||
NM_178221.2 | NP_835739.1 | 11 | 1981 | |||
XM_005271288.2 | XP_005271345.1 | 11 | 1972 | |||
XM_017002600.1 | XP_016858089.1 | 10 | 1883 | |||
XM_017002601.1 | XP_016858090.1 | 10 | 1861 | |||
GenBank mRNA | AK027773.1 | - | 11 | 1999 | ||
BM720875.1 | - | 1 | 346 | |||
BQ777265.1 | - | 1 | 209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
