siRNA Details
Gene Symbol: MGC16025 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026664.1 | 1 | 1379 | |||
GenBank mRNA | BC008026.1 | - | 1 | 1379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
