siRNA Details
Gene Symbol: TMEM128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297551.1 | NP_001284480.1 | 5 | 580 | ||
NM_001297552.1 | NP_001284481.1 | 5 | 578 | |||
NM_032927.3 | NP_116316.1 | 5 | 1085 | |||
XM_005248034.2 | XP_005248091.1 | 5 | 1424 | |||
GenBank mRNA | AK001453.1 | - | 1 | 230 | ||
BC007729.2 | - | 5 | 1050 | 4303698 | ||
BC091482.1 | - | 5 | 580 | 30344390 | ||
BM263799.1 | - | 5 | 560 | |||
CR933611.1 | - | 3 | 4524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
