siRNA Details
Gene Symbol: PNPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033109.4 | NP_149100.2 | 4 | 423 | ||
XM_005264629.2 | XP_005264686.1 | 4 | 353 | |||
XM_017005172.1 | XP_016860661.1 | 3 | 303 | |||
GenBank mRNA | AJ458465.1 | - | 4 | 354 | ||
AK307589.1 | - | 353 | ||||
AY027528.1 | - | 4 | 350 | |||
BC053660.1 | - | 4 | 337 | 6062060 | ||
DA998523.1 | - | 4 | 389 | |||
DB461021.1 | - | 4 | 423 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
