siRNA Details
Gene Symbol: SIGLEC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171156.1 | NP_001164627.1 | 2 | 563 | ||
NM_001171157.1 | NP_001164628.1 | 2 | 563 | |||
NM_001171158.1 | NP_001164629.1 | 2 | 563 | |||
NM_001171159.1 | NP_001164630.1 | 2 | 563 | |||
NM_001171161.1 | NP_001164632.1 | 2 | 563 | |||
NM_001322105.1 | NP_001309034.1 | 2 | 563 | |||
NM_033130.4 | NP_149121.2 | 2 | 563 | |||
XM_005259366.4 | XP_005259423.1 | 2 | 391 | |||
XM_005259367.3 | XP_005259424.1 | 2 | 396 | |||
XM_011527439.1 | XP_011525741.1 | 2 | 391 | |||
GenBank mRNA | AF301007.1 | - | 2 | 458 | ||
AF310233.1 | - | 2 | 378 | |||
AF311905.1 | - | 2 | 563 | |||
AK122619.1 | - | 1 | 273 | |||
AK289900.1 | - | 2 | 349 | |||
AK290542.1 | - | 2 | 396 | |||
AK292025.1 | - | 2 | 466 | |||
AK294185.1 | - | 2 | 402 | |||
AK303514.1 | - | 2 | 402 | |||
AK303717.1 | - | 2 | 402 | |||
AY032685.1 | - | 2 | 497 | |||
AY358337.1 | - | 2 | 386 | |||
BC101725.1 | - | 2 | 428 | 8069231 | ||
BC143497.1 | - | 2 | 428 | 9052005 | ||
BC143500.1 | - | 2 | 428 | 9052008 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
