siRNA Details
Gene Symbol: ROS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002944.2 | NP_002935.2 | 4 | 472 | ||
XM_006715548.3 | XP_006715611.1 | 5 | 606 | |||
XM_011536049.2 | XP_011534351.1 | 5 | 607 | |||
XM_011536050.2 | XP_011534352.1 | 5 | 607 | |||
XM_011536051.2 | XP_011534353.1 | 4 | 580 | |||
XM_011536052.2 | XP_011534354.1 | 5 | 607 | |||
XM_011536053.2 | XP_011534355.1 | 5 | 607 | |||
XM_011536054.2 | XP_011534356.1 | 5 | 608 | |||
XM_011536055.2 | XP_011534357.1 | 5 | 607 | |||
XM_011536056.2 | XP_011534358.1 | 5 | 606 | |||
XM_011536057.2 | XP_011534359.1 | 5 | 605 | |||
XM_011536058.2 | XP_011534360.1 | 5 | 605 | |||
XM_017011172.1 | XP_016866661.1 | 4 | 580 | |||
XM_017011173.1 | XP_016866662.1 | 4 | 682 | |||
GenBank mRNA | BC144556.1 | - | 4 | 320 | ||
M34353.1 | - | 4 | 479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
