siRNA Details
Gene Symbol: ZNF658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317916.1 | NP_001304845.1 | 5 | 2259 | ||
NM_033160.6 | NP_149350.3 | 5 | 2102 | |||
XM_005272515.4 | XP_005272572.1 | 5 | 2209 | |||
XM_011545679.2 | XP_011543981.1 | 5 | 2033 | |||
XM_017014614.1 | XP_016870103.1 | 4 | 4395 | |||
GenBank mRNA | AK022636.1 | - | 1 | 949 | ||
AK024222.1 | - | 1 | 897 | |||
AK292150.1 | - | 5 | 2241 | |||
AL110217.1 | - | 1 | 872 |
Gene Symbol: ZNF658B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003528.3 | 5 | 2111 | |||
GenBank mRNA | AK095694.1 | - | 1 | 1458 | ||
BC137089.1 | - | 1 | 1458 | 9021086 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
