siRNA Details
Gene Symbol: LDHAL6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033195.2 | NP_149972.1 | 1 | 1114 | ||
GenBank mRNA | AK058192.1 | - | 1 | 1044 | ||
AK312777.1 | - | 1 | 1064 | |||
AY009108.1 | - | 1 | 1087 | |||
AY642121.1 | - | 1 | 1107 | |||
BC022034.1 | - | 1 | 1100 | 4825988 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
