siRNA Details
Gene Symbol: DMKN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033746.1 | 13, 14 | 722 | |||
XM_011527495.2 | XP_011525797.1 | 22, 23 | 1855 | |||
XM_011527498.2 | XP_011525800.1 | 21, 22 | 1813 | |||
GenBank mRNA | AK096215.1 | - | 11, 12 | 1261 | ||
BC011886.2 | - | 13, 14 | 651 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
