siRNA Details
Gene Symbol: TRIM55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033058.2 | NP_149047.2 | 8 | 1308 | ||
NM_184085.1 | NP_908973.1 | 8 | 1308 | |||
NM_184086.1 | NP_908974.1 | 8 | 1308 | |||
XM_005251316.4 | XP_005251373.1 | 10 | 2330 | |||
XM_011517613.2 | XP_011515915.1 | 10 | 2333 | |||
XM_011517614.2 | XP_011515916.1 | 10 | 2256 | |||
XM_011517615.2 | XP_011515917.1 | 10 | 2333 | |||
XM_011517616.2 | XP_011515918.1 | 10 | 2333 | |||
XM_011517617.2 | XP_011515919.1 | 8 | 1390 | |||
XM_017013908.1 | XP_016869397.1 | 8 | 1026 | |||
XM_017013909.1 | XP_016869398.1 | 7 | 845 | |||
GenBank mRNA | AJ243488.1 | - | 8 | 1308 | ||
AJ243489.1 | - | 8 | 1308 | |||
AJ291712.1 | - | 8 | 1214 | |||
AJ431704.1 | - | 8 | 1308 | |||
AK091310.1 | - | 8 | 1458 | |||
AK091728.1 | - | 8 | 1128 | |||
BC007750.2 | - | 8 | 1214 | 4110783 | ||
BT007212.1 | - | 8 | 1034 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
