siRNA Details
Gene Symbol: PSIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128217.2 | NP_001121689.1 | 16 | 2997 | ||
NM_033222.4 | NP_150091.2 | 16 | 3000 | |||
GenBank mRNA | AF063020.1 | - | 16 | 2966 | ||
AF432220.1 | - | 16 | 2948 | |||
U94319.1 | - | 10 | 2120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
