siRNA Details
Gene Symbol: SLC25A51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033412.3 | NP_219480.1 | 3 | 617 | ||
GenBank mRNA | BC008500.1 | - | 3 | 378 | 4286914 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
