siRNA Details
Gene Symbol: ARMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199196.1 | NP_001186125.1 | 5 | 1253 | ||
NM_033415.3 | NP_219483.1 | 4 | 1145 | |||
XM_005260157.3 | XP_005260214.1 | 4 | 1196 | |||
XM_017027484.1 | XP_016882973.1 | 4 | 1120 | |||
XM_017027485.1 | XP_016882974.1 | 6 | 1377 | |||
GenBank mRNA | AK295488.1 | - | 4 | 919 | ||
AK303914.1 | - | 2 | 833 | |||
BC056161.1 | - | 6 | 875 | |||
BC067125.1 | - | 5 | 1022 | 6161112 | ||
BX648486.1 | - | 4 | 1040 | |||
CR749678.1 | - | 1 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
