siRNA Details
Gene Symbol: ITPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324240.1 | NP_001311169.1 | 1, 2 | 266 | ||
NM_033453.3 | NP_258412.1 | 1, 2 | 266 | |||
NR_052002.1 | 1, 2 | 266 | ||||
XM_006723564.3 | XP_006723627.1 | 1, 2 | 549 | |||
XM_011529234.2 | XP_011527536.1 | 1, 2 | 549 | |||
GenBank mRNA | AB062127.1 | - | 1, 2 | 135 | ||
AF026816.2 | - | 1, 2 | 125 | |||
AF063607.1 | - | 1, 2 | 150 | |||
AF219116.1 | - | 1, 2 | 178 | |||
BC010138.2 | - | 1, 2 | 110 | 4124624 | ||
BM560812.1 | - | 266 | ||||
BQ049090.1 | - | 1, 2 | 113 | |||
EF199841.1 | - | 1, 2 | 113 | |||
KU178148.1 | - | 1, 2 | 74 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
