siRNA Details
Gene Symbol: ZNF257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316996.1 | NP_001303925.1 | 5 | 1508 | ||
NM_001316997.1 | NP_001303926.1 | 5 | 1452 | |||
NM_001316998.1 | NP_001303927.1 | 2 | 1164 | |||
NM_033468.3 | NP_258429.2 | 4 | 1387 | |||
GenBank mRNA | AF070651.1 | - | 5 | 1204 | ||
AK056694.1 | - | 4 | 1272 | |||
BC036446.1 | - | 5 | 1502 | 4831021 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
