siRNA Details
Gene Symbol: ZNF273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC007078.1 | - | 5 | 1334 | ||
BC019234.1 | - | 5 | 1338 | |||
BC108740.1 | - | 4 | 1052 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
