siRNA Details
Gene Symbol: STARD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243466.1 | NP_001230395.1 | 5 | 537 | ||
NM_001243474.1 | NP_001230403.1 | 5 | 567 | |||
NM_001243476.2 | NP_001230405.1 | 9 | 977 | |||
NM_052851.2 | NP_443083.1 | 5 | 567 | |||
NM_178006.3 | NP_821074.1 | 5 | 630 | |||
NM_178007.2 | NP_821075.1 | 5 | 537 | |||
XM_011535299.2 | XP_011533601.1 | 8 | 723 | |||
XM_017020834.1 | XP_016876323.1 | 5 | 735 | |||
XM_017020835.1 | XP_016876324.1 | 7 | 656 | |||
GenBank mRNA | AK091804.1 | - | 5 | 539 | ||
AK308430.1 | - | 5 | 627 | |||
AK308453.1 | - | 5 | 676 | |||
AK312888.1 | - | 5 | 623 | |||
AL049801.1 | - | 5 | 523 | |||
AY082589.1 | - | 5 | 625 | |||
AY082590.1 | - | 5 | 549 | |||
AY082591.1 | - | 5 | 523 | |||
AY366448.1 | - | 5 | 565 | |||
BC046563.1 | - | 5 | 538 | 5527069 | ||
BP233049.1 | - | 5 | 568 | |||
BX647695.1 | - | 9 | 937 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
