siRNA Details
Gene Symbol: YIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039671.2 | NP_001034760.1 | 8 | 1138 | ||
NM_001039672.2 | NP_001034761.1 | 8 | 1156 | |||
NM_001039673.2 | NP_001034762.1 | 8 | 1147 | |||
NM_001145461.1 | NP_001138933.1 | 8 | 1132 | |||
NM_001145462.1 | NP_001138934.1 | 8 | 1108 | |||
NM_001145463.1 | NP_001138935.1 | 9 | 1224 | |||
XM_005259385.3 | XP_005259442.1 | 8 | 1369 | |||
XM_017027450.1 | XP_016882939.1 | 9 | 1232 | |||
XM_017027451.1 | XP_016882940.1 | 9 | 1262 | |||
GenBank mRNA | AF258578.3 | - | 7 | 1330 | ||
AL833382.1 | - | 9 | 1181 | |||
AY358965.1 | - | 7 | 1275 | |||
BC007644.1 | - | 8 | 1095 | |||
BC091477.1 | - | 8 | 1130 | 30344388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
