siRNA Details
Gene Symbol: MAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052886.2 | NP_443118.1 | 4 | 1071 | ||
XM_011516807.2 | XP_011515109.1 | 3 | 855 | |||
GenBank mRNA | AK172820.1 | - | 4 | 988 | ||
AL117612.1 | - | 1 | 362 | |||
AY007723.1 | - | 4 | 1048 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
