siRNA Details
Gene Symbol: STARD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243476.2 | NP_001230405.1 | 18 | 5501 | ||
NM_052851.2 | NP_443083.1 | 14 | 5091 | |||
NM_178006.3 | NP_821074.1 | 14 | 5154 | |||
NM_178007.2 | NP_821075.1 | 14 | 5061 | |||
XM_011535299.2 | XP_011533601.1 | 17 | 5247 | |||
XM_017020834.1 | XP_016876323.1 | 14 | 5259 | |||
XM_017020835.1 | XP_016876324.1 | 16 | 5180 | |||
GenBank mRNA | AL049801.1 | - | 14 | 5047 | ||
AY082589.1 | - | 14 | 5149 | |||
AY082590.1 | - | 14 | 5073 | |||
AY082591.1 | - | 14 | 5047 | |||
BX641149.1 | - | 4 | 2174 | |||
BX647695.1 | - | 18 | 5461 | |||
CB163673.1 | - | 1 | 410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
