siRNA Details
Gene Symbol: MARVELD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052858.5 | NP_443090.4 | 3 | 1496 | ||
GenBank mRNA | BC011893.2 | - | 3 | 1463 | 3942185 | |
BC013376.2 | - | 3 | 1463 | 3946889 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
