siRNA Details
Gene Symbol: RFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052859.3 | NP_443091.1 | 11, 12 | 1270 | ||
XM_011534216.2 | XP_011532518.1 | 11, 12 | 1368 | |||
XM_017007460.1 | XP_016862949.1 | 11, 12 | 1282 | |||
XM_017007461.1 | XP_016862950.1 | 12, 13 | 1454 | |||
GenBank mRNA | AJ318099.1 | - | 11, 12 | 1254 | ||
BC006846.1 | - | 2, 3 | 184 | |||
BC043595.1 | - | 11, 12 | 1263 | 5769824 |
